Would you like to log in again?
Whatman™ Quartz Air Sampling Filter, Grade QM-A, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
1851-150 1851-050 1851-055 1851-037 1851-085 1851-085 1851-110 1851-037 1851-101 1851-045 1851-050 1851-055 1851-045 1851-047 1851-865 1851-865 1851-8866 1851-025 1851-8866 1851-032 1851-118 1851-025 1851-090 1851-150 1851-082 1851-118 1851-101 1851-110 1851-032 1851-090 1851-047 1851-082
28495-783PK 1927.55 USD InStock
28495-783 CA11008-590 28495-781 11008-560 89187-090 CA70250-028 10035-330 28495-624 28495-646 28495-602 89187-092 89187-094 89187-096 10035-332 CA11013-208 CA11013-668 CA11013-666 10147-920 CA28495-781 11008-590 CA89187-096 CA11008-560 70250-028 CA89187-090 CA89187-092 CA89187-094 CA28495-783 CA10147-920 10035-328 CA28495-624 CA28495-602 CA28495-646
Whatman™ Quartz Air Sampling Filter, Grade QM-A, Whatman products (Cytiva)
Filters Glass Fiber Filters
These high-purity quartz microfiber filters are used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Filters are suitable for PM-10 sampling and analysis.
  • Binder-free
  • High loading capacity
Because of the low level of alkaline earth metals, 'artifact' products of sulfates and nitrates (from SO₂ and NO₂) are virtually eliminated. High-purity quartz microfiber filters are therefore applicable for particulate monitoring and heavy metal analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty