Would you like to log in again?
Whatman™ Grade 934-AH Glass Microfiber Filters, Binder Free
Supplier: Whatman products (Cytiva)
Whatman™ 934-AH™
1827-808 1827-024 1827-032 1827-110 1827-957 1827-185 1827-085 1827-042 1827-320 1827-240 1827-037 1827-150 1827-028 1827-037 1827-105 1827-957 1827-042 1827-021 1827-025 1827-055 1827-028 1827-070 1827-125 1827-070 1827-047 1827-030 1827-185 1827-110 1827-035 1827-090 1827-808 1827-021 1827-035 1827-085 1827-082 1827-047 1827-125 1827-150 1827-866 1827-320 1827-866 1827-889 1827-030 1827-024 1827-055 1827-090 1827-082 1827-032 1827-025 1827-889 1827-240 1827-105
28496-966PK 292.18 USD InStock
CA28496-853 CA28496-897 28496-966 28496-886 CA28496-970 CA28496-773 28297-422 28496-809 CA28496-978 28496-967 28496-968 CA28496-933 CA10147-918 CA28496-809 CA14232-152 CA28496-886 28496-955 28496-911 28496-875 28496-795 28496-751 CA28297-422 CA28496-967 CA28496-968 CA28496-966 28297-418 CA28497-991 CA28496-875 CA28297-418 28496-785 CA28496-751 CA28496-795 CA28496-911 CA28496-955 28497-991 97016-986 28496-780 28496-933 28496-853 CA28496-785 28496-897 CA11013-148 CA11013-146 14232-152 28496-970 28496-773 28496-978 10147-918 CA97016-986 10035-326 CA28496-780 10035-324
Whatman™ Grade 934-AH Glass Microfiber Filters, Binder Free
Filters Glass Fiber Filters
These borosilicate glass filters are specified for water pollution monitoring, cell harvesting, liquid scintillation counting and air pollution monitoring. They are often used in total suspended solids analysis.
  • Smooth surface for efficient precipitate recovery
  • High loading capacity
  • Binder free
Total suspended solids analysis (previously called non-filterable residue) requires high loading capacity glass microfiber filters, such as grade 934-AH.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty