Would you like to log in again?
Whatman™ Grade GF/D Glass Microfiber Binder Free, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
1823-024 1823-915 1823-007 1823-035 1823-055 1823-915 1823-090 1823-257 1823-042 1823-024 1823-047 1823-035 1823-150 1823-047 1823-042 1823-257 1823-010 1823-090 1823-007 1823-021 1823-142 1823-010 1823-025 1823-025 1823-110 1823-125 1823-150 1823-070 1823-021 1823-125 1823-142 1823-110
10147-896PK 81.94 USD InStock
CA28297-416 CA11013-138 CA11013-950 CA11013-136 CA70240-134 10147-896 28497-890 28161-403 28161-500 CA28161-498 CA28161-495 CA28161-491 10035-316 CA11013-946 CA28497-823 CA28497-867 CA28497-801 CA28497-725 70240-134 10035-318 CA28161-403 CA28161-500 28161-498 10035-320 28161-495 28161-491 28497-889 28497-867 28497-823 28497-801 10035-322 28497-725
Whatman™ Grade GF/D Glass Microfiber Binder Free, Whatman products (Cytiva)
Filters Glass Fiber Filters
These borosilicate glass filters are fine membrane filters that can also function as a prefilter when used with GF/B.
  • Thick filter media
  • Good loading capacity
  • Fit most filter holders
  • Binder free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty