Would you like to log in again?
Whatman™ Grade GF/F Glass Microfiber Filters, Binder Free
Supplier: Whatman products (Cytiva)
Whatman™
1825-037 1825-037 1825-024 1825-915 1825-024 1825-070 1825-047 1825-021 1825-257 1825-055 1825-110 1825-110 1825-257 1825-042 1825-142 1825-015 1825-090 1825-293 1825-185 1825-915 1825-150 1825-015 1825-047 1825-150 1825-125 1825-025 1825-293 1825-142 1825-070 1825-090 1825-185 1825-055 1825-025 1825-125
10147-898PK 1150.47 USD InStock
10147-898 28497-909 76635-524 28497-973 CA10147-916 28497-970 CA76205-424 28497-972 CA28497-969 CA28497-925 CA10147-898 CA28497-905 28497-911 28497-936 28497-914 28497-958 CA28497-907 CA28497-909 CA76635-525 CA28497-972 CA28497-973 76205-424 CA28497-970 CA28497-936 CA28497-914 CA28497-958 28497-905 28497-907 10147-916 CA28497-911 28497-903 28497-969 28497-947 28497-925
Whatman™ Grade GF/F Glass Microfiber Filters, Binder Free
Filters Glass Fiber Filters
These borosilicate glass filters are ideal for measuring landfill leachate composition with a TCLP test. They can also be used for DNA binding and purification, filtering finely precipitated proteins and filtering biochemical solutions, fluids, and nucleic acids.
  • Very fine particle retention
  • Good loading capacity and flow rate
  • Binder free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty