Would you like to log in again?
VWR® Talon® Open Rings
Supplier: VWR
VWR® Talon®
80862635 80862637 80862636
28641-014EA 81.24 USD InStock
28641-014 28641-012 28641-010
VWR® Talon® Open Rings
Support Rings
Lightweight rings mount funnels, boiling flasks, and other irregular-shaped objects to lab frames.
  • Aluminum construction
  • Lightweight
  • Easy passing of items through support area
Open ring section allows items to pass in and out of support area easily, reducing risk of breakage. Mount to rods and frames with connectors such as VWR® Talon® regular or heavy-duty holders (see 21572-501 and -566). Extension arm length: 57 mm (21/4").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty