Would you like to log in again?
CASE MINI S BLACK
Supplier: PELICAN PRODUCTS
1200 Case^ Interior 9.25x7.12x4.12in. Watertight, crushproof, dust proof. Double throw latches, Open cell core with solid wall design - strong, light weight. Features an O-ring seal, Automatic Pressure Equalization Valve, Stainless steel hardware, Pick 'N' Pluck* with convoluted lid foam. Black.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CASE MINI S BLACK 28800-004 1200-000-110 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty