Would you like to log in again?
CASE KING PELICAN SILVER
Supplier: PELICAN PRODUCTS
1600 Case^ Interior: 21.43x 16.50x 7.87in. Watertight, crushproof, dust proof. Open cell core with solid wall design - strong, light weight. Automatic Pressure Equalization Valve, O-ring seal, rubber over-molded handle and SS hardware. 2 level Pick 'N' Pluck* with convoluted lid foam. Silver.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CASE KING PELICAN SILVER 28800-038 1600-000-180 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty