Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Transwell® Dish Inserts, Polycarbonate, Sterile, Corning
Supplier: Corning
Corning®
3420
29442-116CS 617.02 USD InStock
29442-116
Transwell® Dish Inserts, Polycarbonate, Sterile, Corning
Culture Dishes Cell Culture Dishes
These tissue culture treated polycarbonate inserts are for use in epithelial and endothelial cell culture, transport, migration/invasion, co-culture, and microbial pathogenesis studies
Inserts provide independent access to both the apical and basolateral sides of the cell monolayer.
Ordering information: Inserts are supplied with dishes. Individually wrapped.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty