Would you like to log in again?
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
Corning® Transwell®
3421 7910 3402 3428 3412 3415 3422 3414 3413 3401
29442-118CS 362.24 USD InStock
29442-118 76404-722 29442-108 29442-106 29442-104 29442-088 29442-110 29442-130 29442-086 29442-120
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Microplates Insert Plates and Inserts
75 mm Transwell® with 0.4 µm pore polycarbonate membrane insert, sterile.
  • 10 µm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Each insert is packaged in a 100 mm dish, 12 inserts per case
  • Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty