Would you like to log in again?
Corning® Cell Lifter and Scrapers, Sterile, Corning
Supplier: Corning
Corning®
3011 3010 3008
29442-204CS 439.01 USD InStock
29442-204 29442-202 29442-200
Corning® Cell Lifter and Scrapers, Sterile, Corning
Cell Scrapers
Useful for the manual harvesting of cells, scrapers are designed for use in flasks and cell lifters for use in harvesting cells (especially stem cells) in dishes.
  • Blade design minimizes cell damage and ensures even contact with the growth surface
  • Individually wrapped
  • Sterilized by gamma radiation
  • Certified non-pyrogenic
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty