Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Costar® Spin-X® Centrifuge Tube Filters, Corning
Supplier: Corning
Corning®
8162 8160 8170 8169 9301 8161 8163
29442-756CS 363.96 USD InStock
29442-756 29442-754 29442-752 29442-762 29442-760 33500-692 29442-758
Costar® Spin-X® Centrifuge Tube Filters, Corning
Filters Centrifugal Filters
Spin-X® centrifuge tube filters use centrifugation to remove bacteria, particles, or cells from liquids, HPLC sample preparation and DNA removal from agarose or acrylamide gels.
  • Cellulose acetate (CA) membranes provide fast flow rates and low protein binding
  • Sterile and nonpyrogenic and DNase-/RNase-free
  • Maximum relative centrifugal force is 16000×g
Units consist of a membrane-containing insert and a polypropylene centrifuge tube. Choose the cellulose acetate membrane insert for aqueous solutions or the nylon membrane insert for HPLC applications. Both membranes are available in 0.22 and 0.45 µm pore sizes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty