Would you like to log in again?
Corning® Gel-Loading Pipette Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
4815 4884 4853 4854
29442-668CS 163.09 USD InStock
29442-668 29442-666 29443-316 29443-288
Corning® Gel-Loading Pipette Tips, Corning
Pipette Tips
These gel-loading pipette tips feature a capillary end that allows easy access into vertical and horizontal electrophoresis gels.
  • Disposable
  • Certified RNase-/DNase-free and non pyrogenic
Ordering information: Available in flat and round tip configurations. Flat tips are designed for use in 0.2 and 0.4 mm sequencing gels. Round tips are designed for use in SDS-PAGE 0.5 mm gels.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty