Would you like to log in again?
Corning® Pipette Tips, Microvolume
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
4834 4830 4901 4894 4840 4826
29443-302CS 100.89 USD InStock
29443-302 29443-304 29443-320 29443-298 29443-300 29443-322
Corning® Pipette Tips, Microvolume
Pipette Tips
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
  • All racked tips are certified RNase-/DNase-free and non pyrogenic
  • Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty