Would you like to log in again?
Stripwell™ 96-well Standard Polystyrene Microplates, Corning®
Supplier: Corning
Corning®
3913 2593 2580 3924 2592 3923 3914 9102
29444-024CS 937.12 USD InStock
29444-024 29442-304 29444-028 29442-302 10068-004 29444-026 29444-030 29442-072
Stripwell™ 96-well Standard Polystyrene Microplates, Corning®
Microplates Immunology Microplates and Strips
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an 'egg-crate' style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • 1×8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty