Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fluid A Rinsing Solution, Biomerieux
Supplier: bioMérieux
P1240-SC P1730 302150 33871 P3140 42616 42044 P1240-SC 42044 42616 302154
95061-460CS 232.79 USD InStock
95061-460 CA95060-750 95061-544 95060-780 95060-750 95061-442 CA-95061-442 29446-137 95060-728 29446-135 CA-95060-780
Fluid A Rinsing Solution, Biomerieux
Microbiology Test Systems Endotoxin Tests
Sterile diluent for rinsing membranes during sterility testing.
  • Effective
  • Large quantity of solution
Rinses membranes during sterility testing by membrane filtration on articles containing lecithin or oil recommended by USP/EP/JP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty