Would you like to log in again?
Performance Low Pressure Air/Water Hose Reel, Coxreels®, ORS Nasco
Supplier: ORS Nasco
170-P-LP-350
300000-083EA 442.2 USD InStock
300000-083
Performance Low Pressure Air/Water Hose Reel, Coxreels®, ORS Nasco
Pneumatic Tools
Easily wraps, stores, and protects hoses to improve efficiency. Rolled edges and ribbed discs for maximum strength and protection. Multi-position locking ratchet secures hose at desired length. Factory-matched cartridge-style lubricated spring motor.
  • Brass NPT swivel inlet machined from solid
  • Multi-position locking ratchet secures hose at desired length
  • Guide arm adjusts for wall, ceiling, or floor mounting
  • Hose Length: 50'
  • Hose Size: 0.375" I.D., 0.65625" O.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty