Would you like to log in again?
Saber Tooth Ceramic Flap Discs, Weiler®
Supplier: Weiler
804-50106 804-50101 804-50107 804-50102 804-50105 804-50103
300000-925EA 15.85 USD InStock
300000-925 300000-926 300000-927 300000-928 300000-923 300000-924
Saber Tooth Ceramic Flap Discs, Weiler®
Abrasives
  • Fast cut and long life due to the aggressive self sharpening ceramic grain
  • Specially formulated grinding aid prevents dulling and glazing of grain and allows for "cool cutting" protecting high value parts from heat discoloration
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty