Would you like to log in again?
Cold Weld Compounds, J-B Weld
Supplier: J-B Weld
803-8265-S
300001-045EA 18.06 USD InStock
300001-045
Cold Weld Compounds, J-B Weld
Adhesives
This is an easy, convenient, and inexpensive alternative to welding, soldering, and brazing.
  • Can be drilled, tapped, machined, filed, sanded and painted
  • Superstrong and nontoxic
  • Stress resistance: 7320 psi
  • Tensile strength: 3960 psi
Will bond to virtually any clean, dry surface including: iron, steel, copper, aluminum, brass, bronze, pewter, porcelain, ceramic, marble, glass, concrete, fiberglass, wood, plastic and many more.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty