Would you like to log in again?
Series 293 Coolant Proof Micrometers, Mitutoyo
Supplier: Mitutoyo
504-293-340
300001-064EA 397.25 USD InStock
300001-064
Series 293 Coolant Proof Micrometers, Mitutoyo
Micrometers
Micrometer with two measuring modes: ABS (absolute) and INC (incremental), for efficient measurement
  • Extra large LCD
  • Carbide-tipped measuring faces
  • Origin setting function (ABS mode): The origin point value can be displayed with a touch of the ORIGIN button
  • Zero setting function (INC mode)
IP65 Dust/Water protection level
Ordering information: Supplied with battery and carrying case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty