Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
62P Series Small Diameter Clamps, Ideal®
Supplier: Ideal®
420-62P10 420-6204 420-62P08 420-62P16 420-6206
300001-218PK 88.33 USD InStock
300001-218 300001-221 300001-223 300001-213 300001-224
62P Series Small Diameter Clamps, Ideal®
Clamps General Purpose Clamps
Suitable for use in such small hose applications as emission control, fuel lines and vacuum hoses.
  • Tiny micro-gear clamps are tough and precision-engineered
  • Slim width is ideal for small or confined area installation
  • Stainless steel band
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty