Would you like to log in again?
Tungsten Carbide Tip Scribers, General Tools
Supplier: General Tools
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
318-88
300002-328EA 14.25 USD InStock
300002-328
Tungsten Carbide Tip Scribers, General Tools
Pens Stylus and Scribers
Free line scriber with straight point.
  • Interchangeable point
  • With pocket clip
For use on ceramics, copper, glass or stainless steel.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty