Would you like to log in again?
Torqueplus™ 12-Point Combination Wrench Sets, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1200FASD 577-1200F-T500
300002-704PK 777.17 USD InStock
300002-704 300023-532
Torqueplus™ 12-Point Combination Wrench Sets, Proto®, ORS Nasco
Wrenches
Increased torque applied to fastener with fewer rounded edges.
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Reduces risk of fastener rounding
  • Box end is precision broached
  • Improves grip on worn, rounded fasteners
  • Applies more torque to fastener
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty