Would you like to log in again?
Torqueplus™ 12-Point Metric Combination Wrenches - Satin Finish, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1215MASD 577-1229MASD 577-1211MASD 577-1217MASD 577-1223MASD 577-1216MASD 577-1212MASD 577-1209MA 577-1232MASD 577-1206MA 577-1241M 577-1210MASD 577-1220MASD 577-1207MA 577-1230MASD 577-1222MASD 577-1250M 577-1227MASD 577-1214MASD 577-1226MASD 577-1224MASD 577-1228MASD 577-1236M 577-1219MASD 577-1208MA 577-1218MASD 577-1221MASD 577-1225MASD 577-1246M 577-1213MASD
300002-708EA 27.56 USD InStock
300002-708 300023-790 300023-780 300023-660 300023-760 300023-684 300023-740 300023-786 300023-764 300023-568 300023-602 300023-768 300023-724 300023-706 300023-580 300023-792 300023-650 300023-672 300002-734 300002-712 300023-750 300023-696 300023-556 300023-734 300023-558 300023-756 300023-638 300023-716 300023-618 300002-730
Torqueplus™ 12-Point Metric Combination Wrenches - Satin Finish, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty