Would you like to log in again?
Proto® Aligning Pry Bars, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-2124 577-2116 577-2125 577-2120
300002-798EA 32.18 USD InStock
300002-798 300002-800 300002-802 300002-804
Proto® Aligning Pry Bars, ORS Nasco
Crow Bars
These black oxide pry bars are used for prying, positioning, and aligning applications.
  • Provides both chisel and tapered tips for a wide range of tasks
  • Durable hardened tool steel construction
  • Chisel - Straight; Point - Straight Tapered Tip Type
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty