Would you like to log in again?
Proto® Metric L-Key Sets, Hex Ball Tip, Metric Stanley
Supplier: Stanley
577-4996
300002-938PK 53.39 USD InStock
300002-938
Proto® Metric L-Key Sets, Hex Ball Tip, Metric Stanley
Wrenches
Proto® Metric ball-hex L-key sets.
  • Rust-resistant black oxide finish
  • Secure fastener fit
  • Heat treated
Delivery information: Set includes 9 Hex keys per holder of different tip sizes 0.15 cm, 0.2 cm, 0.25 cm, 0.3 cm, 0.4 cm, 0.5 cm, 0.6 cm, 0.8 cm, 0.1 cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty