Would you like to log in again?
Proto® Screw Extractors, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-9525B 577-9528 577-9525 577-9529 577-9527 577-9523 577-9526 577-9524 577-9521
300003-212EA 20.72 USD InStock
300003-212 300003-224 300003-210 300003-222 300003-220 300003-218 300003-208 300003-216 300003-214
Proto® Screw Extractors, ORS Nasco
Screwdrivers
Proto® Screw extractors with size stamped on extractor refers to drill bit size to be used with that extractor.
  • Drive into the end of broken pipes or fittings
  • Use tap holder or wrench on extractor to turn out the broken part
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty