Would you like to log in again?
Plastic Pipe and Tubing Cutters, Ridgid®, ORS Nasco
Supplier: ORS Nasco
632-23493
300003-498EA 75.15 USD InStock
300003-498
Plastic Pipe and Tubing Cutters, Ridgid®, ORS Nasco
Cutters
Cutters with X-CEL™ blade for fast single stroke cutting, no twisting or ratcheting required.
  • Durable construction for heavy duty use
Applicable on PVC, CPVC, PEX, PE, PB and Rubber Hose.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty