Would you like to log in again?
Instant Change™ Utility Knives, Stanley®
Supplier: Stanley®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Instant Change™
680-10-788
300003-880EA 34.64 USD InStock
300003-880
Instant Change™ Utility Knives, Stanley®
Knives
Easy blade change and slide operation for quick and easy loading.
  • Push button, quick blade change - no tools required
  • Strong and durable construction with a cast metal core and bi-material soft panels for better grip
  • Curved handle design for better comfort, control and balance
  • Swing out blade storage for easy blade access
  • Integrated string cutting feature
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty