Would you like to log in again?
Wire Rope Cutters, Knipex, ORS Nasco
Supplier: ORS Nasco
414-9561190
300004-820EA 156.25 USD InStock
300004-820
Wire Rope Cutters, Knipex, ORS Nasco
Cutters
Polished cutters with sharp hardened (61 HRC) precision ground cutting edges, factory tested.
  • Angular blades designed for clean cutting of wire, ropes and steel cables
  • Has an opening spring and latch to aid in repetitive use
  • Plastic coated handle
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty