Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tongue and Groove Pliers, Channellock®, ORS Nasco
Supplier: ORS Nasco
140-424-BULK 140-430-BULK 140-460-BULK 140-440-BULK 140-420-BULK
300004-926EA 26.26 USD InStock
300004-926 300004-928 300005-478 300005-476 300004-934
Tongue and Groove Pliers, Channellock®, ORS Nasco
Pliers
Pliers features V-jaws gives more point of contact on round stock and tubing.
  • Patented undercut tongue and groove design assures, no slip jaw action
  • Patented reinforcing edge minimizes stress breakage
  • Patented PermaLock® fastener eliminates nut and bolt failures
  • With polished finish and plastic dipped handle
Features right angle teeth grip in all directions for maximum bite and minimum wear.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty