Would you like to log in again?
Self-Retracting Utility Knife, Stanley®
Supplier: Stanley®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
680-10-189C
300005-602EA 22.81 USD InStock
300005-602
Self-Retracting Utility Knife, Stanley®
Knives
Spring-loaded blade retracts into knife automatically when pressure on slide button is released
Knife can't be left around or put in a pocket with the blade exposed. INTERLOCK® nose secures blade safely between knife halves.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty