Would you like to log in again?
3M™ Industrial Scotch Box Sealing Tapes, 3M Company
Supplier: 3M
5011132205
300005-776EA 32.6 USD InStock
300005-776
3M™ Industrial Scotch Box Sealing Tapes, 3M Company
Tapes
Scotch box sealing tapes seals well in damp conditions (40 to 50 °F), and on recycled fiberboard.
  • Acrylic adhesive offers instant seal
  • Polypropylene backing adds tear resistance and excellent aging
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty