Would you like to log in again?
Rubber Straps, Keeper®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
130-06235
300006-908PK 24.56 USD InStock
300006-908
Rubber Straps, Keeper®, ORS Nasco
Ties
  • Compounded EPDM rubber will not crack, rot or lose rebound qualities
  • It is low temperature resistant - nonbrittle to -40 °F
  • Strong hooks will not bend, break or rust under normal use
  • End type: Steel Hooks
  • Resistant to corrosive, sunlight, ozone, weather
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty