Would you like to log in again?
Regular Face Shields, Crews
Supplier: Crews
135-181540
300006-917EA 7.98 USD InStock
300006-917
Regular Face Shields, Crews
Face Shields
General purpose clear face shield lens made from PC
  • Impact protection
  • Maximum visibility
Dimensions: 8” x 15.5”
Thickness: 0.04"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty