Would you like to log in again?
Plastic Steel® 5 Minute® Putty (SF), Devcon
Supplier: Devcon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Plastic Steel® 5 Minute®
230-10240
300007-389EA 201.09 USD InStock
300007-389
Plastic Steel® 5 Minute® Putty (SF), Devcon
Adhesives
Fast-curing, steel-filled epoxy for emergency repairs at temperatures as low as 40°F.
  • Fast curing
  • Cures to a tough, steel-filled material
  • 5-minute working time
Can be machined, drilled, tapped, or sanded.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty