Would you like to log in again?
Brass Cylinder Adaptors, Western Enterprises
Supplier: Western Enterprises
312-321 312-415 312-830 312-858 312-15 312-831 312-810 312-802 312-813 312-416 312-315 312-320 312-318 312-322 312-809 312-828 312-317 312-316 312-61 312-304 312-16 312-820 312-812 312-51 312-811 312-815 312-808 312-319 312-324 312-305 312-314 312-843 312-827 312-806 312-306
300066-466EA 38.79 USD InStock
300066-466 300007-904 300007-805 300007-828 300066-468 300007-869 300007-902 300066-462 300007-903 300087-804 300066-322 300066-470 300066-472 300007-841 300007-840 300066-456 300066-478 300007-838 300066-458 300007-839 300066-452 300007-836 300007-859 300066-454 300007-834 300066-475 300007-835 300007-832 300066-480 300007-833 300007-797 300007-852 300007-830 300066-460 300007-850
Brass Cylinder Adaptors, Western Enterprises
Welding Supplies
Use a brass cylinder adaptor to connect a regulator to a cylinder or other equipment when different CGA threads on the mating parts make a direct connection impossible.
Adaptors should not be used as permanent installations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty