Would you like to log in again?
Brass Hose Splicers, Western Enterprises
Supplier: Western Enterprises
312-34 312-47 312-53 312-44 312-39 312-57 312-48 312-46 312-55 312-149 312-37 312-54 312-35 312-38 312-45 312-D-38
300066-768EA 68.37 USD InStock
300066-768 300066-356 300066-378 300066-334 300066-336 300066-254 300007-847 300066-366 300007-856 300007-857 300066-332 300007-843 300007-854 300066-392 300007-855 300007-860
Brass Hose Splicers, Western Enterprises
Welding Supplies
  • Made from brass to ensure functionality and durability
  • Can accommodate up to 200 psi of pressure
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty