Would you like to log in again?
Brass Hose Ferrules, Western Enterprises
Supplier: Western Enterprises
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
312-769 312-7331 312-7329 312-620 312-624 312-7333 312-7332 312-7322 312-7330-B 312-5029-A 312-4750 312-7327 312-7328 312-7325 312-7323 312-625 312-3588 312-7326 312-626 312-K-K
300066-414EA 0.51 USD InStock
300066-414 300066-446 300007-858 300007-900 300007-846 300007-898 300007-899 300007-896 300066-362 300007-874 300007-897 300007-875 300007-985 300007-894 300007-873 300007-895 300007-892 300007-893 300007-890 300007-891
Brass Hose Ferrules, Western Enterprises
Welding Supplies
Made from brass to ensure functionality and durability.
Brass hose ferrules provide fast and semi-permanent attachment of hose barbs and most low pressure general purpose hoses.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty