Would you like to log in again?
Inert Arc Nuts, Brass, Western Enterprises
Supplier: Western Enterprises
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
312-AW-14A 312-AW-14 312-AW-21A
300066-514EA 244.41 USD InStock
300066-514 300007-913 300007-914
Inert Arc Nuts, Brass, Western Enterprises
Welding Supplies
  • Made from brass to ensure functionality and durability
  • Can accommodate up to 200 psi of pressure
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty