Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hotspotter All Purpose Propane Torch, Western Enterprises

Supplier: Western Enterprises
312-WB-102 312-WB-101
300008-022EA 125.81 USD InStock
300008-022 300008-021
Hotspotter All Purpose Propane Torch, Western Enterprises
Welding Supplies
Flame tools are for exterior maintenance and repair. The Hotspotter is an all-purpose propane torch that delivers at full cylinder pressure with no smoke and no fumes.

  • Ergonomic 31" handle with adjustable trigger
  • Hooks up to standard 20 lb. propane tank
  • Output Power: 550,000 Btu/h

Heavy Duty torch includes brass propane tank adaptor, brass flow control shut-off valve, spark ignitor, cast iron and steel nozzle, and neoprene hose.

Standard duty torch includes brass propane tank adaptor, brass flow control shut-off valve, spark ignitor, welded tube nozzle, and neoprene hose.

Certifications: Meets NFPA 51 Specifications; CSA Approved.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty