Would you like to log in again?
Flowmeters/Regulators, Gentec
Supplier: Gentec
331-191AR-60
300008-122EA 196.98 USD InStock
300008-122
Flowmeters/Regulators, Gentec
Welding Supplies
  • Underwriters Laboratories Inc. listed
  • Single stage construction
  • Sintered bronze inlet filter
  • Chrome plated housing cap
  • Easily read high-pressure gauge
  • Internal safety pressure relief design
  • Calibrated for argon, helium and carbon dioxide gases
  • Polycarbonate flow tube and outer cover are impact resistant for maximum durability and service life
  • 0-70 SCFH Argon flow, 0-150 SCFH Helium, 0-60 SCFH Carbon Dioxide flow
  • Polycarbonate flow tube and outer cover are impact resistant for longer life
  • Flowmeter is back pressure compensated to ensure accurate reading when line restrictions are present
  • 2" high pressure gauge for easy reading
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty