Would you like to log in again?
Microtorch with Refillable Fuel Cell, Master Appliance, ORS Nasco
Supplier: ORS Nasco
467-MT-11
300009-157EA 15.11 USD InStock
300009-157
Microtorch with Refillable Fuel Cell, Master Appliance, ORS Nasco
Soldering Supplies Soldering Irons
Unit dimensions are 4.3" x 2.5" L x 1" W.
  • 2500 °F operating temperature
  • Butane gas
  • Adjustable pinpoint flame
  • 20 minute run time
Meets CPSC requirements.
Certifications: Meets CPSC Requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty