Would you like to log in again?
SPC™ Oil Sorbents, SPC
Supplier: SPC
SPC™
655-SPC100
300009-857CS 253.88 USD InStock
300009-857
SPC™ Oil Sorbents, SPC
Spill Control Materials
  • Designed to contain and control petroleum based fluids
  • They will not deteriorate, disintegrate, or sink, extruded from polypropylene
  • Heavy weight, dimpled and perfed
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty