Would you like to log in again?
Protektive Pak® ESD Storage Containers, Desco Industries
Supplier: Desco Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
37514 37517 37512 37516
30001-280EA 49.45 USD InStock
30001-280 30001-180 30001-260 30000-986
Protektive Pak® ESD Storage Containers, Desco Industries
Boxes ESD-Safe Boxes
Provide ESD and physical protection for ESD-sensitive components and devices
The conductive plastic handles provide easier handling of the container, as well as a reliable path-to-ground. The steel wire reinforcing frame provides additional strength and durability.
Complete Faraday cage protection. Containers are made of durable fluted conductive cardboard and feature an impregnated liner. Buried shielding layer of <104ohms/sq. minimizes sloughing and rub-off contamination. Feature a static-dissipative surface of 107–109ohms/sq. These reusable containers are constructed with double sides, triple ends, and a double thickness bottom for durability. Made from 100% recycled material and are 100% recyclable.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty