Would you like to log in again?
Weiler® Stem-Mounted Wide Conflex Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-17619 804-17620 804-17623 804-17601 804-17604 804-17621 804-17603 804-17608 804-17602 804-17637 804-17622
300048-140EA 25.77 USD InStock
300048-140 300048-460 300046-302 300046-016 300046-020 300048-144 300010-190 300046-934 300048-136 300048-138 300046-018
Weiler® Stem-Mounted Wide Conflex Brush, ORS Nasco
Brushes
Designed for light duty applications. Use to remove rust, paint, oxides, and scale
  • Wide face
  • Covers a broader surface area
Can be used with power drills, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty