Would you like to log in again?
Back-up Pad for Plastic Button Style Blending, Weiler®
Supplier: Weiler
804-51551
300010-281EA 32.54 USD InStock
300010-281
Back-up Pad for Plastic Button Style Blending, Weiler®
Abrasives
Compatible with 3M Roloc™ back-up pads
Use with right angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty