Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Weiler Tiger Al-tra CUT Discs, Weiler®
Supplier: Weiler
804-59865 804-59862 804-59823 804-59821 804-59824 804-59864
300046-372CS 179.37 USD InStock
300046-372 300047-474 300042-598 300010-293 300048-976 300047-848
Weiler Tiger Al-tra CUT Discs, Weiler®
Abrasives
  • Specially formulated for superior grinding performance on aluminum
  • Resist loading, ensuring higher cut rates and consistent contact with work surface
  • Built for endurance
  • Suitable for aluminum applications such as weld blending, grinding, deburring and finishing
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty