Would you like to log in again?
Small Hand Scratch Brushes, Weiler®
Supplier: Weiler
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
804-95013 804-44637 804-44189 804-95014
300010-255EA 639.65 USD InStock
300010-255 300049-148 300010-295 300010-296
Small Hand Scratch Brushes, Weiler®
Brushes
For general cleaning in the plating, metal finishing, aircraft and missile industries
  • Sanded hardwood block finish
Also used with inert arc aluminum welding.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty