Would you like to log in again?
Small Hand Scratch Brushes, Weiler®
Supplier: Weiler®
804-99383 804-44619 804-44075 804-44167
300010-254EA 38.29 USD InStock
300010-254 300010-238 300048-754 300010-297
Small Hand Scratch Brushes, Weiler®
Brushes
Designed for general cleaning in the plating, metal finishing, aircraft, and missile industries.
Also suitable for use with inert arc aluminum welding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty