Would you like to log in again?
Spatulas, Ampco Safety Tools
Supplier: Ampco Safety Tools
065-S-92 065-S-90 065-S-91
300012-716EA 37.66 USD InStock
300012-716 300012-718 300012-720
Spatulas, Ampco Safety Tools
Spatulas
These spatulas are made of high strength nickel tin copper blade material.
  • Non-sparking
  • Non-magnetic
  • Corrosion resistant
  • Beryllium-free
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty